Spex ISOT-LI6 Claritas PPT® Grade Lithium 6, 100 µg/mL in 2% HNO3; 125 mL

Representative image only.

Spex ISOT-LI6 Claritas PPT® Grade Lithium 6, 100 µg/mL in 2% HNO3; 125 mL

Spex | Item: EW-04502-18

Isotope standards for ICP-MS analysis

$224.00USD / Each

$224.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • For isotope dilution analysis and internal standards
  • 100 ppm lithium 6 in 2% HNO3 matrix
  • 125 mL bottle

More About this Item

The internal standard element must have similar characteristics to the tested/measured element(s) and not be present in the sample. Using isotope modification standards, the chemist can use less internal standard and have a higher intensity reading while avoiding interferences.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Purity GradeClaritas PPT® Grade
  • Concentration100 µg/mL (100 ppm)
  • Matrix2% HNO3
  • Element/IonLithium 6 (Li)
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolIrritant
  • UN NumberUN3264
  • Shelf Life (months)12
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • Polarity Index9
  • Solubility in Water (% wt/wt)100
  • Specific Gravity1
  • UV Cutoff (nm)200
  • DescriptionClaritas PPT® Grade Lithium 6, 100 µg/mL in 2% HNO3; 125 mL
Documents & Certificates
Reviews
Q&A