Spex Lanthanum Solution, APHA, 5% w/v; 20 L

Representative image only.

Spex Lanthanum Solution, APHA, 5% w/v; 20 L

Spex | Item: EW-84410-41

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality

$1,237.00USD / Each

$1,237.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • 20 L Cubitainer® container

More About this Item

Density: 1.1
Composition: water 84.81%, hydrochloric Acid 9.89%, lanthanum Oxide 5.30%
Solubility: miscible
Color: colorless liquid
Packaging: Cubitainer® container
Synonyms: lanthanum solution

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous shipping fee may apply.

Specifications
  • Concentration5% w/v
  • ChemistrySolvents
  • Element/IonLanthanum
  • FormLiquid
  • Volume (mL)20000
  • PackagingCubitainer® container
  • DOT InformationHazardous; Hydrochloric Acid Solution; Class 8, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolCorrosive
  • UN NumberUN1789
  • Shelf Life (months)24
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • DescriptionLanthanum Solution, APHA, 5% w/v; 20 L
Documents & Certificates
Reviews
Q&A