Spex Methanol, ACS Grade, Absolute; 60 L

Representative image only.

Spex Methanol, ACS Grade, Absolute; 60 L

Spex | Item: EW-84410-44

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality

$280.00USD / Each

$280.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • ACS grade—meets or exceeds purity standards set by the American Chemical Society
  • 60 L round metal can

More About this Item

Flash point: 11 °C
Density: 0.79
Molecular weight: 32.0419 g/mol
Solubility: miscible
Refractive index: 1.328 to 1.33nD
Color: colorless liquid

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous shipping fee may apply.

Specifications
  • Purity GradeACS
  • ChemistrySolvents
  • Element/IonMethanol
  • FormLiquid
  • Volume (mL)60000
  • PackagingRound metal can
  • DOT InformationHazardous; Methanol; Class 3 (6.1), Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • CAS Number67-56-1
  • UN NumberUN1230
  • Shelf Life (months)60
  • Boiling Point C (° C)64.5
  • Boiling Point F (° F)148.1
  • Melting Point C (° C)-98
  • Melting Point F (° F)-144.4
  • DescriptionMethanol, ACS Grade, Absolute; 60 L
Documents & Certificates
Reviews
Q&A