Spex Nitric Acid, 70% w/w, Trace Metal, ACS Grade; 2.5 L

Representative image only.

Spex Nitric Acid, 70% w/w, Trace Metal, ACS Grade; 2.5 L

Spex | Item: EW-84410-47

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality

$144.00USD / Each

$144.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • ACS grade—meets or exceeds purity standards set by the American Chemical Society
  • 2.5 L clear glass bottle

More About this Item

Assay percentage: 68.00 to 70.00%
Density: 1.416
Molecular weight: 63.01 g/mol
Solubility: miscible
pH range: < 1
Color: colorless to light yellow liquid

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous shipping fee may apply.

Specifications
  • Purity GradeTrace metals, ACS
  • ChemistrySolvents
  • Element/IonNitric Acid
  • FormLiquid
  • SynonymsAqua fortis, spirit of niter, eau forte, hydrogen nitrate, acidum nitricum
  • Volume (mL)2500
  • PackagingClear glass bottle
  • DOT InformationHazardous; Nitric Acid; Class 8 (5.1), Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolOxidizer, Corrosive
  • CAS Number7697-37-2
  • UN NumberUN2031
  • Shelf Life (months)36
  • Boiling Point C (° C)121
  • Boiling Point F (° F)249.8
  • Melting Point C (° C)-32
  • Melting Point F (° F)-25.6
  • DescriptionNitric Acid, 70% w/w, Trace Metal, ACS Grade; 2.5 L
Documents & Certificates
Reviews
Q&A