Spex Phosphoric Acid, 50% v/v; 4 L

Representative image only.

Spex Phosphoric Acid, 50% v/v; 4 L

Spex | Item: UX-84410-53

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality

$162.00USD / Each

$162.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • 50% v/v concentration
  • 4 L natural poly bottle

More About this Item

Solubility: miscible
pH range: < 1
Color: colorless liquid

Density: 1.36
Molecular weight: 98 g/mol
Composition: water 46.60%, phosphoric acid 53.40%

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous shipping fee may apply.

Specifications
  • Concentration50% v/v
  • ChemistrySolvents
  • Element/IonPhosphoric Acid
  • FormLiquid
  • Volume (mL)4000
  • PackagingNatural poly bottle
  • DOT InformationHazardous; PHOSPHORIC ACID SOLUTION; Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosive
  • CAS Number7664-38-2
  • UN NumberUN1805
  • Shelf Life (months)36
  • Boiling Point C (° C)108
  • Boiling Point F (° F)226.4
  • Melting Point C (° C)-43.6
  • Melting Point F (° F)-42
  • DescriptionPhosphoric Acid, 50% v/v; 4 L
Documents & Certificates
Reviews
Q&A