Spex CertiPrep® Phosphoric Acid

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality
  • Designed to work with any instrument and application
Solubility: miscible
pH range: < 1
Color: colorless liquid
$162.00 - $265.00USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Concentration Packaging Volume (mL)
Availability Pricing
COMPARE
0.85 Amber glass bottle 4000
$265.00USD / Each
COMPARE
50% v/v Natural poly bottle 4000
$162.00USD / Each
COMPARE
60% v/v Natural poly bottle 4000
$187.00USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat