Spex PLCA2-2M Assurance® Grade Calcium, 1,000 µg/mL (1,000 ppm) for AA and ICP in 2% HNO3, 30 mL

Representative image only.

Spex PLCA2-2M Assurance® Grade Calcium, 1,000 µg/mL (1,000 ppm) for AA and ICP in 2% HNO3, 30 mL

Spex | Item: EW-04504-65

Single-element calcium standards for AA & ICP analysis

$39.00USD / Each

$39.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Made with acid and ASTM Type I water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity (where commercially available)
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • 30 mL bottle

More About this Item

Assurance® grade certified reference materials (CRMs) are designed for atomic absorption (AA) and inductively coupled plasma (ICP) analysis. They are packaged in 30 mL, 125 mL, 250 mL, and 500 mL bottles to minimize contamination. Custom standards can be manufactured upon request.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Causes skin irritation. Causes serious eye irritation. Wear protective gloves. Wear eye protection/face protection. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Specific treatment (see on this label). If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention.

Irritant
Specifications
  • Purity GradeAssurance® Grade
  • Concentration1,000 µg/mL Ca in 2% HNO3
  • Empirical FormulaCa
  • Element/IonCalcium
  • Volume (mL)30
  • DOT InformationDoes Not Apply
  • Hazard SymbolIrritant
  • UN NumberUN3264
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionCalcium, 1,000 µg/mL (1,000 ppm) for AA and ICP in 2% HNO3, 30 mL
Documents & Certificates
Reviews
Q&A