Spex Potassium Iodide, 10% w/v with 0.3% Sodium Bicarbonate; 4 L

Representative image only.

Spex Potassium Iodide, 10% w/v with 0.3% Sodium Bicarbonate; 4 L

Spex | Item: EW-84410-56

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality

$201.00USD / Each

$201.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • 4 L amber glass bottle

More About this Item

Density: 1.08
Composition: potassium iodide 9.30%, water 90.42%, sodium bicarbonate 0.28%
Solubility: miscible
pH range: 6 to 9.2
Color: colorless to pale yellow liquid

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Concentration10% w/v
  • ChemistrySolvents
  • Element/IonPotassium Iodide
  • FormLiquid
  • Volume (mL)4000
  • PackagingAmber glass bottle
  • DOT InformationNonhazardous; Not Regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • UN NumberNot Regulated
  • Shelf Life (months)18
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)32
  • Melting Point F (° F)0
  • DescriptionPotassium Iodide, 10% w/v with 0.3% Sodium Bicarbonate; 4 L
Documents & Certificates
Reviews
Q&A