Spex Potassium Iodide, ACS Grade; 12 kg

Representative image only.

Spex Potassium Iodide, ACS Grade; 12 kg

Spex | Item: UX-84410-59

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality

$4,092.00USD / Each

$4,092.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • ACS grade—meets or exceeds purity standards set by the American Chemical Society
  • 12 kg poly pail

More About this Item

Density: 3.12
Molecular weight: 166 g/mol
Chemical formula: KI
Composition: potassium iodide 100%
Solubility: 1480 g/L at 25 °C
pH range: 6 to 9.2
Color: white solid

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS
  • Linear FormulaKI
  • ChemistrySolvents
  • Element/IonPotassium Iodide
  • FormSolid
  • Weight (g)12000
  • PackagingPoly pail
  • DOT InformationNonhazardous; Not Regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • CAS Number7681-11-0
  • UN NumberNot Regulated
  • Shelf Life (months)85
  • Boiling Point C (° C)1323
  • Boiling Point F (° F)2413.4
  • Melting Point C (° C)1257.8
  • Melting Point F (° F)681
  • DescriptionPotassium Iodide, ACS Grade; 12 kg
Documents & Certificates
Reviews
Q&A