Spex CertiPrep® Potassium Iodide, ACS Grade

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality
  • Designed to work with any instrument and application
Density: 3.12
Molecular weight: 166 g/mol
Chemical formula: KI
Composition: potassium iodide 100%
Solubility: 1480 g/L at 25 °C
pH range: 6 to 9.2
Color: white solid
$438.00 - $4,092.00USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Form Packaging Description
Availability Pricing
COMPARE
Solid Poly pail Potassium Iodide, ACS Grade; 12 kg
$4,092.00USD / Each
COMPARE
Solid Wide mouth container Potassium Iodide, ACS Grade; 500 mL
$438.00USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat