Spex S-990 Crotonaldehyde Organic Standard, 1,000 µg/mL in DI Water; 1 mL

Representative image only.

Spex S-990 Crotonaldehyde Organic Standard, 1,000 µg/mL in DI Water; 1 mL

Spex | Item: EW-04545-18

Single-component organic standards ensure consistent results

$54.00USD / Each

$54.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm crotonaldehyde single-component organic standard in DI water matrix
  • 1 mL ampule

More About this Item

Single-component standards offer high-quality, consistent results while saving preparation time. 2-Chloronaphthalene standard is supplied in methanol-P&T. Additional solvents include methylene chloride, acetone, benzene, hexane, acetonitrile, THF, and water. Please inquire about the matrix for a particular item when ordering, if the matrix is important to your application, a substitution may be possible.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixDI Water
  • Element/IonCrotonaldehyde
  • FormLiquid
  • Volume (mL)1
  • DOT InformationNot Regulated
  • Storage SymbolRefrigerator
  • Hazard SymbolNot Regulated
  • CAS Number123-73-9
  • UN NumberNot Regulated
  • Shelf Life (months)3
  • DescriptionCrotonaldehyde Organic Standard, 1,000 µg/mL in DI Water; 1 mL
Documents & Certificates
Reviews
Q&A