Spex Sodium Thiosulfate, 0.1 N; 20 L

Representative image only.

Spex Sodium Thiosulfate, 0.1 N; 20 L

Spex | Item: EW-84410-85

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality

$208.00USD / Each

$208.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • 20 L Cubitainer® container

More About this Item

Density: 1.01
Composition: water 97.40%, sodium thiosulfate pentahydrate 2.46%, proprietary 0.09%, sodium carbonate 0.05%
Solubility: miscible
pH range: 6 to 8.4
Color: colorless liquid

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This product is only available in the US.; for a quote, please contact [email protected].

Specifications
  • Concentration0.1 N
  • ChemistrySolvents
  • Element/IonSodium Thiosulfate
  • FormLiquid
  • Volume (mL)20000
  • PackagingCubitainer® container
  • DOT InformationNonhazardous; Not Regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • UN NumberNot Regulated
  • Shelf Life (months)24
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionSodium Thiosulfate, 0.1 N; 20 L
Documents & Certificates
Reviews
Q&A