Spex Tributyl Phosphate, 98.5%+, Purified Grade; 1 L

Representative image only.

Spex Tributyl Phosphate, 98.5%+, Purified Grade; 1 L

Spex | Item: UX-84410-90

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality

$218.00USD / Each

$218.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • 1 L natural poly bottle

More About this Item

Assay percentage: 99+%
Flash point: 146 °C
Density: 0.98
Molecular weight: 266.31 g/mol
Chemical formula: (C4H9)3PO4
Composition: tributyl phosphate 100%
Solubility: 0.28 g/L at 25°C
Refractive index: 1.423 to 1.425nD
Color: colorless to slightly yellow liquid

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear Formula(C4H9)3PO4
  • ChemistrySolvents
  • Element/IonTributyl Phosphate
  • FormLiquid
  • Volume (mL)1000
  • PackagingNatural poly bottle
  • DOT InformationNonhazardous; Not Regulated
  • Storage SymbolAmbient
  • Hazard SymbolIrritant, Environment
  • CAS Number126-73-8
  • UN NumberNot Regulated
  • Shelf Life (months)60
  • Boiling Point C (° C)130
  • Boiling Point F (° F)266
  • Melting Point C (° C)-70
  • Melting Point F (° F)-94
  • DescriptionTributyl Phosphate, 98.5%+, Purified Grade; 1 L
Documents & Certificates
Reviews
Q&A