Spex Tributyl Phosphate

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality
  • Designed to work with any instrument and application
Assay percentage: 99+%
Flash point: 146 °C
Density: 0.98
Molecular weight: 266.31 g/mol
Chemical formula: (C4H9)3PO4
Composition: tributyl phosphate 100%
Solubility: 0.28 g/L at 25°C
Refractive index: 1.423 to 1.425nD
Color: colorless to slightly yellow liquid
$218.00 - $363.00USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Form Volume (mL)
Availability Pricing
COMPARE
Liquid 4000
$363.00USD / Each
COMPARE
Liquid 1000
$218.00USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat