Sussman 39123B Replacment Heating Element, 12 kW, 208 VAC

Representative image only.

Sussman 39123B Replacment Heating Element, 12 kW, 208 VAC

Sussman | Mfr: 39123B| Item: EW-07602-21

$827.00USD / Each

$827.00USD / Each
Product Description

Key Features

  • 1 year warranty

More About this Item

Keep your steam generator running smoothly with these parts and accessories.

1 Year Warranty
Specifications
  • Compatible WithSussman Steam Generator with 12 kW, 208 VAC power
  • Power (VAC)208
  • DescriptionReplacment Heating Element, 12 kW, 208 VAC
  • Warranty1 year
Reviews
Q&A