Testo 0554 0191 Wireless Probe Handle with Thermocouple Probe Connection
Representative image only.
Testo 0554 0191 Wireless Probe Handle with Thermocouple Probe Connection
Testo | Mfr: 0554 0191| Item: EW-37852-54
For use with Testo thermohygrometers
$234.00USD / Each
sign in for your price.
$234.00USD / Each
sign in for your price.
Product Description
Key Features
  • Temperature and humidity probes
  • Communication cables
  • Calibration
More About this Item
Replacement parts and accessories for Testo humidity meters.
Specifications
  • Min RH Range (%)0
  • Max RH Range (%)100
  • RH Accuracy±2.5% RH
  • RH Resolution (%)0.1%
  • Min Temperature (° F)-14
  • Max Temperature (° F)140
  • Temperature Accuracy±0.9°F
  • Temperature Resolution0.1°
  • DescriptionWireless Probe Handle with Thermocouple Probe Connection