Thermo Scientific GENESYS™ Integrated Sipper System for GENESYS™ 100 Series and BioMate 160
Representative image only.
Thermo Scientific GENESYS™ Integrated Sipper System for GENESYS™ 100 Series and BioMate 160
Thermo Scientific | Mfr: 840-306900| Item: EW-02654-45
Enhance the versitility of your spectrophotometer
$2,402.00USD / Each
$2,402.00USD / Each
Buy It Together
Current Item
Thermo Scientific GENESYS™ Integrated Sipper System for GENESYS™ 100 Series and BioMate 160
Thermo Scientific GENESYS™ Integrated Sipper System for GENESYS™ 100 Series and BioMate 160
0265445
$2,402.00USD / Each
+ =
Total Price 6492.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Expand your spectrophotometer capacity and capabilities
  • Fully controlled by the instrument's on-board software and powered by external power supply
  • Sip by time or calibrate to sip by volume
  • Versitile system comes with pre-programmed and custom sip sequences
  • Capable of three pump speeds
  • 2 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories including cuvette holders, sipper pumps, and more suitable for use with the Thermo Scientific™ GENESYS™ visible and UV-visible spectrophotometers. Spectrophotometers sold separately.
sipper pump with pump head, sample compartment insert with cuvette holder and goose-neck aspiration tube guide, quartz flow cell with connector tubing, 30 cm of L/S 16 silicone tubing, power supply and cord, and cable.
Available for sale and use in the United States only.
Two year warranty
Specifications
  • Compatible WithGENESYS 140, 150, 180, and 160 BioMate spectrophotometers
  • DescriptionIntegrated Sipper System for GENESYS™ 100 Series and BioMate 160