Thermo Scientific GENESYS™ Long Path Cylindrical Cell Holder for GENESYS™ 100 Series and BioMate 160; 5 to 100 mm
Representative image only.
Thermo Scientific GENESYS™ Long Path Cylindrical Cell Holder for GENESYS™ 100 Series and BioMate 160; 5 to 100 mm
Thermo Scientific | Mfr: 840-303100| Item: EW-02654-34
Enhance the versitility of your spectrophotometer
$312.75USD / Each
$312.75USD / Each
Buy It Together
Current Item
Thermo Scientific GENESYS™ Long Path Cylindrical Cell Holder for GENESYS™ 100 Series and BioMate 160; 5 to 100 mm
Thermo Scientific GENESYS™ Long Path Cylindrical Cell Holder for GENESYS™ 100 Series and BioMate 160; 5 to 100 mm
0265434
$312.75USD / Each
+ =
Total Price 4402.75 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Expand your spectrophotometer capacity and capabilities
  • 2 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories including cuvette holders, sipper pumps, and more suitable for use with the Thermo Scientific™ GENESYS™ visible and UV-visible spectrophotometers. Spectrophotometers sold separately.
Long path cylindrical cell holder is suitable for 5 to 100 mm pathlength cuvettes.
Available for sale and use in the United States only.
Two year warranty
Specifications
  • Compatible WithGENESYS 140, 150, 180, and 160 BioMate spectrophotometers
  • DescriptionLong Path Cylindrical Cell Holder for GENESYS™ 100 Series and BioMate 160; 5 to 100 mm