Thermo Scientific GENESYS™ Single Cell Cuvette Holder and Base for GENESYS™ 100 Series and BioMate 160
Representative image only.
Thermo Scientific GENESYS™ Single Cell Cuvette Holder and Base for GENESYS™ 100 Series and BioMate 160
Thermo Scientific | Mfr: 840-298900| Item: EW-02654-31
Enhance the versitility of your spectrophotometer
$193.25USD / Each
$193.25USD / Each
Buy It Together
Current Item
Thermo Scientific GENESYS™ Single Cell Cuvette Holder and Base for GENESYS™ 100 Series and BioMate 160
Thermo Scientific GENESYS™ Single Cell Cuvette Holder and Base for GENESYS™ 100 Series and BioMate 160
0265431
$193.25USD / Each
+ =
Total Price 4283.25 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Expand your spectrophotometer capacity and capabilities
  • 2 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories including cuvette holders, sipper pumps, and more suitable for use with the Thermo Scientific™ GENESYS™ visible and UV-visible spectrophotometers. Spectrophotometers sold separately.
Standard 10 mm single-cell holder for GENESYS™ 100 Series and BioMate 160 spectrophotometers.
Available for sale and use in the United States only.
Two year warranty
Specifications
  • Compatible WithGENESYS 140, 150, 180, and 160 BioMate spectrophotometers
  • DescriptionSingle Cell Cuvette Holder and Base for GENESYS™ 100 Series and BioMate 160