Thermo Scientific GENESYS™ WiFi USB Adaptor for GENESYS™ 40, 50, 140, 150, 180 and BioMate 160
Representative image only.
Thermo Scientific GENESYS™ WiFi USB Adaptor for GENESYS™ 40, 50, 140, 150, 180 and BioMate 160
Thermo Scientific | Mfr: 840-309900| Item: EW-02654-47
Enhance the versitility of your spectrophotometer
$209.25USD / Each
$209.25USD / Each
Buy It Together
Current Item
Thermo Scientific GENESYS™ WiFi USB Adaptor for GENESYS™ 40, 50, 140, 150, 180 and BioMate 160
Thermo Scientific GENESYS™ WiFi USB Adaptor for GENESYS™ 40, 50, 140, 150, 180 and BioMate 160
0265447
$209.25USD / Each
+ =
Total Price 4299.25 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Expand your spectrophotometer capacity and capabilities
  • 2 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories including cuvette holders, sipper pumps, and more suitable for use with the Thermo Scientific™ GENESYS™ visible and UV-visible spectrophotometers. Spectrophotometers sold separately.
The Wi-Fi USB adapter adds wireless capabilities to your GENESYS™ 40, 50, 140, 150, 180 and BioMate 160 spectophotometers. This enables wireless communication directly to your lab network and printers.
Available for sale and use in the United States only.
Two year warranty
Specifications
  • Compatible WithGENESYS 140, 150, 180, and 160 BioMate spectrophotometers
  • DescriptionWiFi USB Adaptor for GENESYS™ 40, 50, 140, 150, 180 and BioMate 160