Thermo Scientific PR-1 NanoDrop™ Spectrophotometer Reconditioning Kit
Representative image only.
Thermo Scientific PR-1 NanoDrop™ Spectrophotometer Reconditioning Kit
Thermo Scientific | Mfr: CHEM-PR1-KIT| Item: EW-83060-94
Optimize your NanoDrop with these calibration accessories
$198.00USD / Each
$220.00USD / Each
$198.00USD / Each
$220.00USD / Each
Buy It Together
Current Item
Thermo Scientific PR-1 NanoDrop™ Spectrophotometer Reconditioning Kit
Thermo Scientific PR-1 NanoDrop™ Spectrophotometer Reconditioning Kit
8306094
$198.00USD / Each
$220.00USD / Each
+
Thermo Scientific 2000 1-position Spectrophotometer
8306100
$13,943.00USD / Each
=
Total Price 14141.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
one screw cap vial of specially formulated compound and 25 applicators.
Key Features
  • Ensure accuracy of your spectrophotometer with these calibration accessories
More About this Item
Choose from a wide range of calibration accessories to keep your Thermo Scientific™ NanoDrop™ spectrophotometers and fluorometers running accurately and efficiently. Spectrophotometers sold separately.
This kit is useful for reconditioning the sample measurement pedestals on the Thermo Scientific NanoDrop™ spectrophotometers to a hydrophobic state. Kit contains specially formulated polishing compound and applicators.
Available for sale and use in the United States only.
Specifications
  • Compatible WithNanoDrop Spectrophotometers
  • DescriptionPR-1 NanoDrop™ Spectrophotometer Reconditioning Kit