Thermo Scientific R01768 Sabouraud dextrose agar, pH 5.6, 10/pk
Representative image only.
Thermo Scientific R01768 Sabouraud dextrose agar, pH 5.6, 10/pk
Thermo Scientific | Mfr: R01768| Item: EW-14207-94
Stringent quality control ensures high-quality performance as intended!
$136.50USD / Pkg of 10
$136.50USD / Pkg of 10
Product Description
Key Features
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
More About this Item
Choose from a variety of media formulations. Use this culture media for the isolation and differentiation of microorganisms from samples or specimens. Each raw component is pretested before being used, for quality assurance and consistency from lot batch. Intended for lab practice only; not intended for in vitro purposes. Plates have stacking rings which interlock to minimize sliding.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
  • Media Name (No.)SabDex (Sabouraud-Dextrose)
  • DescriptionSabouraud dextrose agar, pH 5.6, 10/pk