Thermo Scientific R455052 Tryptic soy broth (TSB), soybean casein digest, 500g
Representative image only.
Thermo Scientific R455052 Tryptic soy broth (TSB), soybean casein digest, 500g
Thermo Scientific | Mfr: R455052| Item: EW-14206-20
Stringent quality control ensures reliable performance
$221.00USD / Each
$221.00USD / Each
Product Description
Key Features
  • For lab use only, not intended for in vitro purposes
More About this Item
Use this culture media for isolation and differentiation of microorganisms from samples or specimens. Choose from a variety of media formulations. Each raw component is pretested before being used for quality assurance and consistency from lot batch. All animal product components are in compliance with USDA and APHIS regulations. For lab use only, not intended for in vitro purposes.
For the cultivation of a wide variety of nonfastidious microorganisms. Conforms to USP.
Available for sale in the United States only
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
  • Media Name (No.)Tryptic Soy
  • Capacity (g)500
  • DescriptionTryptic soy broth (TSB), soybean casein digest, 500g