ThermoProbe PG-050-SW-SM Replacement Probe Assembly 50ft Std

Representative image only.

ThermoProbe PG-050-SW-SM Replacement Probe Assembly 50ft Std

ThermoProbe | Mfr: PG-050-SW-SM| Item: EW-90026-06

Replacement Probes for Models TP7 and TP9

$508.05USD / Each

$508.05USD / Each
Product Description

Key Features

  • 3 month warranty

More About this Item

Convenient replacement probes for ThermoProbe meters, TP7 and TP9.

3 Month Warranty
Specifications
  • Meter Measurement TypeRTD
  • Number Of Probe Channels1
  • Types Of Probes AcceptedRTD
  • Width (in)600
  • Depth (in)1/4
  • DescriptionReplacement Probe Assembly 50ft Std
  • Warranty3 month
Reviews
Q&A