Trajan SGE Replacement Plunger, Compatible with Agilent Fixed Needle GT Syringe, 10 uL, 23 G; 2/Pk
Representative image only.
Trajan SGE Replacement Plunger, Compatible with Agilent Fixed Needle GT Syringe, 10 uL, 23 G; 2/Pk
Trajan | Mfr: 031808| Item: EW-16002-33
Choose wisely to save your operational cost—no need to buy new syringes instead
$88.00USD / Pkg of 2
$88.00USD / Pkg of 2
Buy It Together
Current Item
Trajan SGE Replacement Plunger, Compatible with Agilent Fixed Needle GT Syringe, 10 uL, 23 G; 2/Pk
Trajan SGE Replacement Plunger, Compatible with Agilent Fixed Needle GT Syringe, 10 uL, 23 G; 2/Pk
1600233
$88.00USD / Pkg of 2
+ =
Total Price 227.25 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Each stainless steel plunger is individually fitted to each barrel
  • 6 month warranty
More About this Item
All SGE gas-tight syringes have replaceable plungers.
The standard plunger in MicroVolume syringes is not replaceable.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Replacement Plunger, Compatible with Agilent Fixed Needle GT Syringe, 10 uL, 23 G; 2/Pk