Trajan SGE Replacement Plunger, Compatible with CTC PAL Systems Gas Tight Syringe, 100 uL
Representative image only.
Trajan SGE Replacement Plunger, Compatible with CTC PAL Systems Gas Tight Syringe, 100 uL
Trajan | Mfr: 0318261| Item: EW-16002-51
Choose wisely to save your operational cost—no need to buy new syringes instead
$53.70USD / Pkg of 1
$53.70USD / Pkg of 1
Buy It Together
Current Item
Trajan SGE Replacement Plunger, Compatible with CTC PAL Systems Gas Tight Syringe, 100 uL
Trajan SGE Replacement Plunger, Compatible with CTC PAL Systems Gas Tight Syringe, 100 uL
1600251
$53.70USD / Pkg of 1
+ =
Total Price 192.95 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Each stainless steel plunger is individually fitted to each barrel
  • 6 month warranty
More About this Item
All SGE gas-tight syringes have replaceable plungers.
The standard plunger in MicroVolume syringes is not replaceable.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Replacement GT plunger for 100 uL Fixed Needle CTC Syringe, 0.47 mm OD, Cone Tip; 1/Ea