Trajan SGE Replacement Plunger, Compatible with CTC/Thermo Gas Tight Syringe, 10 uL, 5 cm L; 2/Pk
Representative image only.
Trajan SGE Replacement Plunger, Compatible with CTC/Thermo Gas Tight Syringe, 10 uL, 5 cm L; 2/Pk
Trajan | Mfr: 0318120| Item: EW-16002-37
Choose wisely to save your operational cost—no need to buy new syringes instead
$79.70USD / Pkg of 2
$79.70USD / Pkg of 2
Buy It Together
Current Item
Trajan SGE Replacement Plunger, Compatible with CTC/Thermo Gas Tight Syringe, 10 uL, 5 cm L; 2/Pk
Trajan SGE Replacement Plunger, Compatible with CTC/Thermo Gas Tight Syringe, 10 uL, 5 cm L; 2/Pk
1600237
$79.70USD / Pkg of 2
+ =
Total Price 218.95 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Each stainless steel plunger is individually fitted to each barrel
  • 6 month warranty
More About this Item
All SGE gas-tight syringes have replaceable plungers.
The standard plunger in MicroVolume syringes is not replaceable.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Replacement Plunger, Compatible with CTC/Thermo Gas Tight Syringe, 10 uL, 5 cm L; 2/Pk