Trajan SGE Replacement Plunger, Compatible with PerkinElmer Gas Tight Syringe, 5 uL
Representative image only.
Trajan SGE Replacement Plunger, Compatible with PerkinElmer Gas Tight Syringe, 5 uL
Trajan | Mfr: 031807| Item: EW-16002-32
Choose wisely to save your operational cost—no need to buy new syringes instead
$77.30USD / Pkg of 1
$77.30USD / Pkg of 1
Buy It Together
Current Item
Trajan SGE Replacement Plunger, Compatible with PerkinElmer Gas Tight Syringe, 5 uL
Trajan SGE Replacement Plunger, Compatible with PerkinElmer Gas Tight Syringe, 5 uL
1600232
$77.30USD / Pkg of 1
+ =
Total Price 216.55 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Each stainless steel plunger is individually fitted to each barrel
  • 6 month warranty
More About this Item
All SGE gas-tight syringes have replaceable plungers.
The standard plunger in MicroVolume syringes is not replaceable.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Replacement Plunger, Compatible with PerkinElmer Gas Tight Syringe, 5 uL; 1/Ea