Trajan SGE Replacement Plunger, Compatible with Varian Autosampler, 10 uL, 5.3 cm, SH Needle
Representative image only.
Trajan SGE Replacement Plunger, Compatible with Varian Autosampler, 10 uL, 5.3 cm, SH Needle
Trajan | Mfr: 031218| Item: EW-16002-14
Choose wisely to save your operational cost—no need to buy new syringes instead
$73.90USD / Each
$73.90USD / Each
Buy It Together
Current Item
Trajan SGE Replacement Plunger, Compatible with Varian Autosampler, 10 uL, 5.3 cm, SH Needle
Trajan SGE Replacement Plunger, Compatible with Varian Autosampler, 10 uL, 5.3 cm, SH Needle
1600214
$73.90USD / Each
+ =
Total Price 213.15 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Each stainless steel plunger is individually fitted to each barrel
  • 6 month warranty
More About this Item
All SGE gas-tight syringes have replaceable plungers.
The standard plunger in MicroVolume syringes is not replaceable.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Replacement Plunger, Compatible with Varian Autosampler, 10 uL, 5.3 cm, SH Needle; 1/Ea