Trajan SGE Replacement Plunger, for 10 uL Fixed Needle Syringe, 50 mm L
Representative image only.
Trajan SGE Replacement Plunger, for 10 uL Fixed Needle Syringe, 50 mm L
Trajan | Mfr: 031803| Item: EW-16002-30
Choose wisely to save your operational cost—no need to buy new syringes instead
$55.40USD / Pkg of 1
$55.40USD / Pkg of 1
Buy It Together
Current Item
Trajan SGE Replacement Plunger, for 10 uL Fixed Needle Syringe, 50 mm L
Trajan SGE Replacement Plunger, for 10 uL Fixed Needle Syringe, 50 mm L
1600230
$55.40USD / Pkg of 1
+ =
Total Price 194.65 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Each stainless steel plunger is individually fitted to each barrel
  • 6 month warranty
More About this Item
All SGE gas-tight syringes have replaceable plungers.
The standard plunger in MicroVolume syringes is not replaceable.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Replacement Plunger, for 10 uL Fixed Needle Syringe, 50 mm L; 1/Ea