Trajan SGE Replacement Plunger, for 10 uL Gas Tight Syringe, Removable Bevel-Tip Needle, 0.47 mm OD
Representative image only.
Trajan SGE Replacement Plunger, for 10 uL Gas Tight Syringe, Removable Bevel-Tip Needle, 0.47 mm OD
Trajan | Mfr: 031811| Item: EW-16002-36
Choose wisely to save your operational cost—no need to buy new syringes instead
$77.40USD / Pkg of 1
$77.40USD / Pkg of 1
Buy It Together
Current Item
Trajan SGE Replacement Plunger, for 10 uL Gas Tight Syringe, Removable Bevel-Tip Needle, 0.47 mm OD
Trajan SGE Replacement Plunger, for 10 uL Gas Tight Syringe, Removable Bevel-Tip Needle, 0.47 mm OD
1600236
$77.40USD / Pkg of 1
+ =
Total Price 216.65 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Each stainless steel plunger is individually fitted to each barrel
  • 6 month warranty
More About this Item
All SGE gas-tight syringes have replaceable plungers.
The standard plunger in MicroVolume syringes is not replaceable.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Replacement GT plunger for 10 uL Removable Needle Syringe, 0.47 mm OD, Bevel Tip; 1/Ea