Trajan SGE Replacement Plunger, for Removable Luer Lock Syringe; 100 mL
Representative image only.
Trajan SGE Replacement Plunger, for Removable Luer Lock Syringe; 100 mL
Trajan | Mfr: 0312176| Item: EW-16002-13
Choose wisely to save your operational cost—no need to buy new syringes instead
$172.75USD / Each
$172.75USD / Each
Buy It Together
Current Item
Trajan SGE Replacement Plunger, for Removable Luer Lock Syringe; 100 mL
Trajan SGE Replacement Plunger, for Removable Luer Lock Syringe; 100 mL
1600213
$172.75USD / Each
+ =
Total Price 312.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Each stainless steel plunger is individually fitted to each barrel
  • 6 month warranty
More About this Item
All SGE gas-tight syringes have replaceable plungers.
The standard plunger in MicroVolume syringes is not replaceable.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Replacement Plunger, for Removable Luer Lock Syringe; 100 mL; 1/Ea