Trajan SGE Replacement Plunger with Bevel-Tip Needle, for 500 uL Fixed Needle Syringe
Representative image only.
Trajan SGE Replacement Plunger with Bevel-Tip Needle, for 500 uL Fixed Needle Syringe
Trajan | Mfr: 031835| Item: EW-16002-60
Choose wisely to save your operational cost—no need to buy new syringes instead
$53.00USD / Pkg of 1
$53.00USD / Pkg of 1
Buy It Together
Current Item
Trajan SGE Replacement Plunger with Bevel-Tip Needle, for 500 uL Fixed Needle Syringe
Trajan SGE Replacement Plunger with Bevel-Tip Needle, for 500 uL Fixed Needle Syringe
1600260
$53.00USD / Pkg of 1
+ =
Total Price 192.25 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Each stainless steel plunger is individually fitted to each barrel
  • 6 month warranty
More About this Item
All SGE gas-tight syringes have replaceable plungers.
The standard plunger in MicroVolume syringes is not replaceable.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Replacement GT plunger with BT Needle, for 500 uL Fixed Needle Syringe; 1/Ea