US Industrial 5200 Analog Gas Detector/Alarm, Ammonia, Wall Mount with built-in sensor

Representative image only.

US Industrial 5200 Analog Gas Detector/Alarm, Ammonia, Wall Mount with built-in sensor

US Industrial | Mfr: 5200| Item: EW-10109-95

Don’t risk workers’ safety

$1,540.00USD / Each

$1,540.00USD / Each
Product Description

Key Features

  • Continuous 24 hour monitoring Compact, durable units Explosion proof housing for sensor Audio/visual alarms Malfunction LED and test/checking switches Infrequent maintenance and calibration required Remote sensor reliable to 600 feet from controller Relay contacts and recorder terminals
  • 1 year warranty

More About this Item

The wall mount unit is for use in an enclosed area. Remote sensor provides open space monitoring ability. At hazardous locations, explosion proof sensors and their junction box housing meet NEC requirements for Class 1, Division 1, Group A-B-C-D. The unit is designed for those who are concerned about explosive and toxic gases over a long period of time mostly within an enclosed area and/or work environment.

1 Year Warranty
Specifications
  • Gases MeasuredAmmonia
  • AlarmAlarm light (red), fail light (yellow), and audible alarm
  • Display TypeAnalog
  • Power (VDC)24
  • Power (VAC)115
  • Height (in)8
  • Width (in)5
  • Depth (in)4
  • Height (cm)20.3
  • Width (cm)12.7
  • Depth (cm)10.2
  • DescriptionAnalog Gas Detector/Alarm, Ammonia, Wall Mount with built-in sensor
  • Warranty1 year
Reviews
Q&A