Velp A00001305 SS Stirring Shaft with Folding Blade
Representative image only.
Velp A00001305 SS Stirring Shaft with Folding Blade
Velp | Mfr: A00001305| Item: EW-50200-81
$173.50USD / Each
$173.50USD / Each
Buy It Together
Current Item
Velp A00001305 SS Stirring Shaft with Folding Blade
Velp A00001305 SS Stirring Shaft with Folding Blade
5020081
$173.50USD / Each
+ + =
Total Price 1968.50 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 2 year warranty
More About this Item
Choose from our comprehensive line of Velp mixer accessories including stirring shafts, support base, and clamps.
The blade automatically falls in line during rotation to generate an axial flow in the container from top towards the bottom. Recommended for stirring in narrow-neck containers.
Two year warrantyConformite Europeenne
Specifications
  • DescriptionSS Stirring Shaft with Folding Blade