Whatman 6717-9504 Polycap TC Filter Capsule, PES, Sterile, 0.45/0.65um, 2200 cm
Representative image only.
Whatman 6717-9504 Polycap TC Filter Capsule, PES, Sterile, 0.45/0.65um, 2200 cm
Whatman | Mfr: 6717-9504| Item: EW-29701-66
Pre-sterilized polyethersulfone (PES) membrane
$278.55USD / Pkg of 1
$278.55USD / Pkg of 1
Product Description
Key Features
  • Select capsules with filling bell for cleanliness assurance during filling
  • 45 day warranty
More About this Item
These capsules contain two layers of polyethersulfone (PES) membrane, which is inherently hydrophilic, low in extractables, and offers excellent flow rates. The membrane also exhibits low protein binding characteristics making it ideal for use with biologically sensitive solutions. The device is supplied presterilized and has a vent to evacuate any air to allow fast filtration. The outlet of the capsule is protected to prevent touch contamination during setup and has standard hose barb connections.
Sterile
Specifications
  • Membrane MaterialPolyethersulfone
  • Pore Size (micron)0.45/0.65
  • Housing MaterialPolypropylene
  • SterileYes
  • Connections1/4" to 3/8" hose barb
  • Effective Filtration Area (cm²)220
  • Surface Area (m²)0.022
  • Max Pressure (PSI)60
  • Max Pressure4.1
  • DescriptionPolycap TC Filter Capsule, PES, Sterile, 0.45/0.65um, 2200 cm
  • Warranty45 days