A&D SJ-WP Series Compact Washdown Bench Scales
Take-Away Check Weighing makes packing from a master container easy and accurate
  • IP67-rated for washdown environments, and submersible up to 3-ft for sink washing
  • 304 stainless steel pan easily removes for cleaning
  • Weighing sensor is sealed and isolated from the electronic components adding durability
  • Traffic light comparator uses red, green, and yellow LED lights and user selectable 3, 5, or 7-stage comparisons to make check weighing quick and precise
  • Tare functions up to full capacity, allowing use of any tare container
  • Automatic power-off feature prolongs battery life
Perfect for food preparation and processing, the delicate internal electronics are isolated from the harsh environment in a washdown submersible housing. Use any container you choose—tare functions up to the scale's full capacity.
The traffic light comparator's 3, 5, and 7-stage comparison function allows you to choose between greater precision and quicker response. Take-away check weighing feature increases flexibility, so you can place a master container on the scale and distribute to smaller containers—reducing costs and providing accurate portion control. With this feature, the scale automatically tares after every successful distribution from the master container.
ITEM(S) ON SALE
$556.10 - $607.50USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Readability (g)
Availability Pricing
A&D Weighing SJ-3000WPF Compact Washdown Scale, 6Lb x 0.0002Lb EW-11115-98 Mfr # SJ-3000WPF
COMPARE
3 6.6 0.1
$571.50USD / Each
$635.00USD / Each
A&D Weighing SJ-6000WPF Compact Washdown Scale, 13Lb x 0.0005Lb EW-11115-99 Mfr # SJ-6000WPF
COMPARE
6 13 0.2
$571.50USD / Each
$635.00USD / Each
A&D Weighing SJ-15KWPF Compact Washdown Scale. 33Lb x 0.0005Lb EW-11116-01 Mfr # SJ-15KWPF
COMPARE
15 33 0.2
$607.50USD / Each
$675.00USD / Each
A&D Weighing SJ-30KWPF Compact Washdown Scale. 66Lb x 0.002Lb EW-11116-02 Mfr # SJ-30KWPF
COMPARE
30 66 1
$556.10USD / Each
$670.00USD / Each