Accelerated One-Step Modular Extractor/Concentrator
Accelerated One-Step™ modular extractor/concentrator performs extraction, concentration, and drying—all in one apparatus! Features PTFE hydrophobic membranes for speedy drying. Meets EPA and CLP standards. Includes extractor cup and body, Snyder sidearm, membrane assembly, and connectors. Order concentrator tube and condenser separately below to fit your specific application.
$454.00USD / Case of 1
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Pyrex 2157-100TJ Jacketed Concentrator Tube 1-Step Modular Glass System, 100 mL EW-34510-02 Mfr # 2157-100TJ
COMPARE
Jacketed Concentrator Tube 1-Step Modular Glass System, 100 mL
$454.00USD / Case of 1