Acros Organics 2,2,4-Trimethylpentane

Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$123.00 - $405.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
Acros Organics 2,2,4-Trimethylpentane, spectrophotometricgrade 99% (2.5l) EW-88178-25 Mfr # AC167910025
COMPARE
2500 2,2,4-Trimethylpentane, spectrophotometricgrade 99% (2.5l)
$405.00USD / Each
Acros Organics 2,2,4-Trimethylpentane, for HPLC (1l) EW-88259-29 Mfr # AC268810010
COMPARE
1000 2,2,4-Trimethylpentane, for HPLC (1l)
$206.75USD / Each
Acros Organics 2,2,4-Trimethylpentane c.p. (2.5l) EW-88271-73 Mfr # AC295720025
COMPARE
2500 2,2,4-Trimethylpentane c.p. (2.5l)
$248.00USD / Each
Acros Organics 2,2,4-Trimethylpentane, reagent ACS (500 mL) EW-88363-37 Mfr # AC424565000
COMPARE
500 2,2,4-Trimethylpentane, reagent ACS (500 mL)
$123.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat