Acros Organics N,N-Dimethylacetamide

Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$99.20 - $223.75USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
CAS Number Volume (mL) Description
Availability Pricing
Acros Organics N,N-Dimethylacetamide 99%; 2.5 L EW-88109-42 Mfr # AC115690025
COMPARE
127-19-5 2500 N,N-Dimethylacetamide 99% (2.5l) CAS 127-19-5
$205.75USD / Each
Acros Organics N,N-Dimethylacetamide, 99+%, Spectrophotometric Grade; 250 mL EW-88178-01 Mfr # AC167782500
COMPARE
127-19-5 250 N,N-Dimethylacetamide, spectrophotometricgrade 99+% (250 mL)
$99.20USD / Each
Acros Organics N,N-Dimethylacetamide, 99+%; 2.5 L EW-88194-41 Mfr # AC181070025
COMPARE
127-19-5 2500 N,N-Dimethylacetamide 99+% (2.5l) CAS 127-19-5
$223.75USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat