Acros Organics Silicone Oil
Improve performance of your lab equipment
  • Reagents and solutions to meet your lab and research needs
Silicone oils have thermal stability, electrical insulation, and lubrication properties. Their temperature stability and good heat-transfer characteristics make them ideal for use in laboratories for heating baths placed on top of hotplate stirrers, as well as in freeze-dryers as refrigerants.
The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$141.00 - $192.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Weight (g) Description
Availability Pricing
Acros Organics Silicone Oil, for Melting Point and Boiling Point Apparatuses; 1 kg EW-88172-31 Mfr # AC163850010
COMPARE
63148-62-9 1000 Silicone oil, for melting point and boiling point apparatuses (1kg)
$192.50USD / Each
Acros Organics Silicone Oil, for Melting Point and Boiling Point Apparatuses; 500 g EW-88172-32 Mfr # AC163855000
COMPARE
63148-62-9 500 Silicone oil, for melting point and boiling point apparatuses (500g)
$141.00USD / Each