Advantec Disposable Sterile Syringe Filters, PTFE Hydrophilic
Housings are specifically designed to minimize sample holdup and maximize recovery
  • Non-pigmented acrylic or polypropylene (PP) housings feature integral filter sealing to avoid the risk of contamination from pigments and adhesives
  • Each filter is clearly marked with membrane pore size
  • Housings have standard luer connectors; 50-mm filters feature 7 to 13.5 mm hose barb connectors
PTFE Hydrophilic Filters are versatile filters characterized by very good chemical resistance. They are compatible with many aqueous and organic solvent mixtures used in HPLC, including acetonitrile/water.
$210.65 - $276.40USD / Pkg of 100
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Pore Size (µm)
Availability Pricing
Advantec 13HP020AN Syringe Filters, 0.2 um Hydrophilic PTFE, 13 mm; 100/Pk EW-81054-12 Mfr # 13HP020AN
COMPARE
13 0.2
$210.65USD / Pkg of 100
Advantec 13HP045AN Syringe Filters, 0.5 um Hydrophilic PTFE, 13 mm; 100/Pk EW-81054-14 Mfr # 13HP045AN
COMPARE
13 0.5
$210.65USD / Pkg of 100
Advantec 25HP020AN Syringe Filters, 0.2 um Hydrophilic PTFE, 25 mm; 100/Pk EW-81054-42 Mfr # 25HP020AN
COMPARE
25 0.2
$276.40USD / Pkg of 100