Advantec MCE Membranes for Asbestos Monitoring
Cleanroom manufacturing ensures minimal contamination and accurate results
Offered in 0.80 µm pore size, these filters are manufactured specifically for industrial hygiene sampling of asbestos fibers. Cutting and packaging operations are carried out under cleanroom conditions to keep non-asbestos fiber contamination to an absolute minimum. Filters may be cleared using the acetone vapors method or the dimethylphthalate diethyloxalate method.
$154.05 - $214.75USD / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material Pore Size (µm)
Availability Pricing
Advantec A080X047A MCE Solid White Filters, 0.8um, 47mm Dia; 100/Pk EW-06639-76 Mfr # A080X047A
COMPARE
47 Mixed Cellulose Ester 0.8
$214.75USD / Pkg of 100
Advantec A080X025B MCE Gridded Filters, 0.8um, 25mm Dia; 100/Pk EW-36230-30 Mfr # A080X025B
COMPARE
25 Mixed Cellulose Ester 0.8
$154.05USD / Pkg of 100