AEMC 6240 Micro-Ohmmeter
  • Reliable low resistance measurements
  • Four-terminal Kelvin resistance measurement eliminates test lead resistance
  • 10A test current at up to 4000µΩ
  • 0.25% basic accuracy
  • 1 micro-ohm resolution
  • Direct reading, easy to operate
  • Six selectable resistance ranges
  • Reverse polarity switch
  • Overload and input fuse protection
  • Manufactured to international safety and environmental standards
  • Automatic decimal point and zeroing
  • Large terminals accept banana plugs and spade lugs
  • Rechargeable NiMH battery with internal charger (110/220V)
  • Includes DataView® software for data storage, real-time display, analysis and report generation
$3,534.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
AC Current Accuracy AC Current Range (mA) AC Current Resolution (mA)
Availability Pricing
AEMC 6240 10-Amp Micro-Ohmmeter, 1µ Ohms to 400 Ohms EW-27443-45 Mfr # 2129.8
COMPARE
10 A to 4000 μΩ
$3,534.00USD / Each