AkroBins Stackable Storage Bins
Sturdy, one-piece construction resists rust, water, and corrosion
  • Autoclave at 250° F (121° C)
  • Stackable
AkroBins are not affected by weak acids and bases. Sterilize in an autoclave at 250° F (121° C). Reinforced design prevents them from spreading when they are stacked or loaded.
$19.40 - $371.50USD / Pkg of 12
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Depth (in) Height (in) Width (in)
Availability Pricing
Akro-Mils 30230 Blue Stackable Bin, 5" High; 12 Bins/PK EW-47151-10 Mfr # 30230 BLUE
COMPARE
10.875 5 5.5
$226.25USD / Pkg of 12
Akro-Mils 30239 BLUE Stackable Bin, 7" High, PACK OF 6 BINS 8.25 in W x 7 in H x 10.75 in D EW-47151-12 Mfr # 30239 BLUE
COMPARE
10.75 7 8.25
$173.75USD / Pkg of 6
Akro-Mils 30250 BLUE Stackable Bin, 7" High, PACK OF 6 BINS (BLUE) EW-47151-14 Mfr # 30250 BLUE
COMPARE
14.75 7 16.5
$301.50USD / Pkg of 6
Akro-Mils 30265 BLUE Stackable Bin, 9" High, PACK OF 6 BINS 8.25 in W x 9 in H x 18 in D EW-47151-16 Mfr # 30265 BLUE
COMPARE
14.75 7 8.25
$371.50USD / Pkg of 6
Akro-Mils 30280 BLUE Stackable Bin, 6" High, PACK OF 4 BINS EW-47151-17 Mfr # 30280 BLUE
COMPARE
20 6 12.375
$156.00USD / Pkg of 4
Akro-Mils 30230 BLUE Stackable Bin, 4 3/4" High 5.5 in W x 5 in H x 10.875 in D EW-47151-20 Mfr # 30230 BLUE
COMPARE
10.875 5 5.5
$19.40USD / Each
Akro-Mils 30282 BLUE Stackable Bin, 12" High, Pack Of 2 Bins EW-47151-21 Mfr # 30282 BLUE
COMPARE
20 12 12.375
$106.75USD / Pkg of 2
Akro-Mils 30239 BLUE Stackable Bin, 7" High, 8.25 in W x 7 in H x 10.75 in D EW-47151-22 Mfr # 30239 BLUE
COMPARE
10.75 7 8.25
$29.50USD / Each
Akro-Mils 30250 BLUE Stackable Bin, 7" High 16.5 in W x 7 in H x 14.75 in D EW-47151-24 Mfr # 30250 BLUE
COMPARE
14.75 7 16.5
$51.20USD / Each
Akro-Mils 30265 Blue Stackable Bin, 9" High 8.25 in W x 9 in H x 18 in D EW-47151-26 Mfr # 30265 BLUE
COMPARE
14.75 7 8.25
$62.40USD / Each
Akro-Mils 30282 BLUE Stackable Bin, 12" High (10 11/16"W x 11 3/8"H x 16 7/8"D) EW-47151-32 Mfr # 30282 BLUE
COMPARE
20 12 12.375
$53.80USD / Each
Showing 1- 11 of 11