Alicat LC-Series Liquid Flow Controllers, CCM
Offers fast, precise measurement of liquid flow rate, pressure, and temperature in real-time
  • No warm-up required
  • Thirty millisecond measurement response time
  • Steady state control 2-100% of full scale
  • NIST-traceable accuracy up to ±2% of full scale
  • IP40 rating
  • Lifetime warranty
These liquid flow controllers are designed for high-performance applications where precise monitoring of fluid parameters is crucial. With a rapid control response time of just 30 milliseconds (ms), the controllers provide real-time measurements of several key fluid properties, including flow rate, pressure, and temperature. This makes it ideal for handling a wide range of liquids, including water, methanol, ethanol, ammonia, dimethyl ether, iso-pentane, thiophene, iso-octane, and heptane.
The controllers feature a user-friendly monochrome display that allows for simultaneous viewing of all measurement data, including flow rate (in cubic centimeters per minute, CCM), pressure, and temperature. A proportional control valve is incorporated into the system, enabling precise control of the liquid flow. The flow only activates once the controller sends a voltage signal to the valve, ensuring that no flow occurs without explicit command.
The units come with convenient bleed screws, enabling you to remove air pockets from the system quickly, ensuring optimal performance. Additionally, the data I/O port allows seamless integration with computer systems, enabling convenient data logging and analysis. This feature ensures flow performance can be continuously monitored and analyzed, making the controller a reliable and versatile tool for various industrial applications requiring highly accurate and stable fluid measurements.
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (SCCM) Accuracy Repeatability
Availability Pricing
Alicat Liquid Flow Controller with Display, 0 - 5 CCM EW-15309-31 Mfr # LC-5CCM-D
COMPARE
5 ±2.0% of full scale ±0.2% of full scale
$2,040.00USD / Each
Alicat Liquid Flow Controller with Display, 0 - 10 CCM EW-15309-32 Mfr # LC-10CCM-D
COMPARE
10 ±2.0% of full scale ±0.2% of full scale
$2,040.00USD / Each
Alicat Liquid Flow Controller with Display, 0 - 20 CCM EW-15309-33 Mfr # LC-20CCM-D
COMPARE
20 ±2.0% of full scale ±0.2% of full scale
$2,040.00USD / Each
Alicat Liquid Flow Controller with Display, 0 - 50 CCM EW-15309-34 Mfr # LC-50CCM-D
COMPARE
50 ±2.0% of full scale ±0.2% of full scale
$2,040.00USD / Each
Alicat Liquid Flow Controller with Display, 0 - 100 CCM EW-15309-35 Mfr # LC-100CCM-D
COMPARE
100 ±2.0% of full scale ±0.2% of full scale
$2,040.00USD / Each
Alicat Liquid Flow Controller with Display, 0 - 200 CCM EW-15309-36 Mfr # LC-200CCM-D
COMPARE
200 ±2.0% of full scale ±0.2% of full scale
$2,040.00USD / Each
Alicat Liquid Flow Controller with Display, 0 - 500 CCM EW-15309-37 Mfr # LC-500CCM-D
COMPARE
500 ±2.0% of full scale ±0.2% of full scale
$2,040.00USD / Each
Showing 1- 7 of 7