Alicat MC-Series Mass Flow Controllers, SCCM
Provides accurate, real-time mass and volumetric flow readings
  • No warm-up required
  • Ninety-eight pre-loaded gas calibrations
  • Ten millisecond measurement response time
  • NIST-traceable calibration certificate included - accuracy up to ±0.5% of reading
  • Limited lifetime warranty
These mass flow controllers, featuring a fast control response time of just 30 milliseconds (ms), offer quick integration into test stands for accurate verification without any warm-up time. They are capable of measuring and controlling flow generated by both positive pressure and suction. Real-time monitoring of flow, pressure, and temperature is available directly on the display screen.
The controllers come with 98 pre-loaded gas calibrations, which can be seamlessly switched while maintaining accuracy. The stand-alone units measure mass flow, volumetric flow, temperature, and absolute pressure, with flow rates displayed in standard cubic centimeters per minute (SCCM). Additional key features include an LCD display with backlight, a data I/O port for logging data to your computer, and flexible mounting options, including upside-down installation.
Please note; power supply and cable sold separately.
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (SCCM) Accuracy Repeatability
Availability Pricing
Alicat Mass Flow Controller with Display, 0 - 0.5 SCCM EW-15308-94 Mfr # MC-0.5SCCM-D
COMPARE
0.5 ±0.8% of reading + 0.2% of full scale ±0.2% of reading + 0.02% of full scale
$2,315.00USD / Each
Alicat Mass Flow Controller with Display, 0 - 1 SCCM EW-15308-95 Mfr # MC-1SCCM-D
COMPARE
1 ±0.8% of reading + 0.2% of full scale ±0.2% of reading + 0.02% of full scale
$2,315.00USD / Each
Alicat Mass Flow Controller with Display, 0 - 2 SCCM EW-15308-96 Mfr # MC-2SCCM-D
COMPARE
2 ±0.8% of reading + 0.2% of full scale ±0.2% of reading + 0.02% of full scale
$2,315.00USD / Each
Alicat Mass Flow Controller with Display, 0 - 5 SCCM EW-15308-97 Mfr # MC-5SCCM-D
COMPARE
5 ±0.8% of reading + 0.2% of full scale ±0.2% of reading + 0.02% of full scale
$1,760.00USD / Each
Alicat Mass Flow Controller with Display, 0 - 10 SCCM EW-15308-98 Mfr # MC-10SCCM-D
COMPARE
10 ±0.6% of reading or ±0.1% of full scale, whichever is greater ±0.1% of reading + 0.02% of full scale
$1,760.00USD / Each
Alicat Mass Flow Controller with Display, 0 - 20 SCCM EW-15308-99 Mfr # MC-20SCCM-D
COMPARE
20 ±0.6% of reading or ±0.1% of full scale, whichever is greater ±0.1% of reading + 0.02% of full scale
$1,760.00USD / Each
Alicat Mass Flow Controller with Display, 0 - 50 SCCM EW-15309-00 Mfr # MC-50SCCM-D
COMPARE
50 ±0.6% of reading or ±0.1% of full scale, whichever is greater ±0.1% of reading + 0.02% of full scale
$1,760.00USD / Each
Alicat Mass Flow Controller with Display, 0 - 100 SCCM EW-15309-01 Mfr # MC-100SCCM-D
COMPARE
100 ±0.6% of reading or ±0.1% of full scale, whichever is greater ±0.1% of reading + 0.02% of full scale
$1,760.00USD / Each
Alicat Mass Flow Controller with Display, 0 - 200 SCCM EW-15309-02 Mfr # MC-200SCCM-D
COMPARE
200 ±0.6% of reading or ±0.1% of full scale, whichever is greater ±0.1% of reading + 0.02% of full scale
$1,760.00USD / Each
Alicat Mass Flow Controller with Display, 0 - 500 SCCM EW-15309-03 Mfr # MC-500SCCM-D
COMPARE
500 ±0.6% of reading or ±0.1% of full scale, whichever is greater ±0.1% of reading + 0.02% of full scale
$1,760.00USD / Each
Showing 1- 10 of 10