Alicat MC-Series Mass Flow Controllers, SLPM
Provides accurate, real-time mass and volumetric flow readings
  • No warm-up required
  • Ninety-eight pre-loaded gas calibrations
  • Ten millisecond measurement response time
  • NIST-traceable calibration certificate included - accuracy up to ±0.5% of reading
  • Lifetime warranty
These mass flow controllers, featuring a fast control response time of just 30 milliseconds (ms), offer quick integration into test stands for accurate verification without any warm-up time. They are capable of measuring and controlling flow generated by both positive pressure and suction. Real-time monitoring of flow, pressure, and temperature is available directly on the display screen.
The controllers come with 98 pre-loaded gas calibrations, which can be seamlessly switched while maintaining accuracy. The stand-alone units measure mass flow, volumetric flow, temperature, and absolute pressure, with flow rates displayed in standard liters per minute (SLPM). Additional key features include an LCD display with backlight, a data I/O port for logging data to your computer, and flexible mounting options, including upside-down installation.
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (LPM) Accuracy Repeatability
Availability Pricing
Alicat Mass Flow Controller with Display, 0 - 1 SLPM EW-15309-04 Mfr # MC-1SLPM-D
COMPARE
1 ±0.6% of reading or ±0.1% of full scale, whichever is greater ±0.1% of reading + 0.02% of full scale
$1,760.00USD / Each
Alicat Mass Flow Controller with Display, 0 - 2 SLPM EW-15309-05 Mfr # MC-2SLPM-D
COMPARE
2 ±0.6% of reading or ±0.1% of full scale, whichever is greater ±0.1% of reading + 0.02% of full scale
$1,815.00USD / Each
Alicat Mass Flow Controller with Display, 0 - 5 SLPM EW-15309-06 Mfr # MC-5SLPM-D
COMPARE
5 ±0.6% of reading or ±0.1% of full scale, whichever is greater ±0.1% of reading + 0.02% of full scale
$1,760.00USD / Each
Alicat Mass Flow Controller with Display, 0 - 10 SLPM EW-15309-07 Mfr # MC-10SLPM-D
COMPARE
10 ±0.6% of reading or ±0.1% of full scale, whichever is greater ±0.1% of reading + 0.02% of full scale
$1,760.00USD / Each
Alicat Mass Flow Controller with Display, 0 - 20 SLPM EW-15309-08 Mfr # MC-20SLPM-D
COMPARE
20 ±0.6% of reading or ±0.1% of full scale, whichever is greater ±0.1% of reading + 0.02% of full scale
$1,760.00USD / Each