Alicat Whisper™ MWB-Series Low-Pressure Drop Mass Flowmeters, Digital Display, Battery Operated, SLPM
Provides accurate real-time mass and volumetric flow readings with minimal pressure drop
  • Instant warm-up
  • Ninety-eight pre-loaded gas calibrations
  • One-hundred millisecond control response
  • Built-in rechargeable battery with up to 18 hours life
  • IP40 rating
These portable, low-pressure drop mass flowmeters utilize laminar differential pressure technology to deliver precise mass flow measurements for over 130 different gases. With a rapid control response time of just 100 milliseconds, they integrate easily into test stands, enabling immediate and accurate verification without the need for warm-up.
Designed to minimize pressure drop—as low as 0.07 psid—these flowmeters are ideal for applications with limited source pressure. Each unit comes pre-loaded with 98 standard gas calibrations that can be easily switched without compromising accuracy.
These stand-alone devices measure mass flow, volumetric flow, temperature, and absolute pressure, with readings shown in standard liters per minute (SLPM). Additional features include a backlit monochrome LCD display, a data I/O port for seamless data logging to a computer, and versatile mounting options—even allowing for upside-down installation.
$1,970.00 - $6,340.00USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max Flow Rate (LPM) Repeatability
Availability Pricing
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter, 0 - 1 SLPM, Battery Operated EW-67803-25 Mfr # MWB-1SLPM-D
COMPARE
±0.75% of reading ±0.1% of full scale 1 ±0.1% of reading ±0.02% of full scale
$1,970.00USD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter, 0 - 2 SLPM, Display, Battery EW-67803-26 Mfr # MWB-2SLPM-D
COMPARE
±0.75% of reading ±0.1% of full scale 2 ±0.1% of reading ±0.02% of full scale
$1,970.00USD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter, 0 - 5 SLPM, Display, Battery EW-67803-27 Mfr # MWB-5SLPM-D
COMPARE
±0.75% of reading ±0.1% of full scale 5 ±0.1% of reading ±0.02% of full scale
$1,970.00USD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter, 0 - 10 SLPM, Display, Battery EW-67803-28 Mfr # MWB-10SLPM-D
COMPARE
±0.75% of reading ±0.1% of full scale 10 ±0.1% of reading ±0.02% of full scale
$1,970.00USD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter, 0 - 20 SLPM, Display, Battery EW-67803-29 Mfr # MWB-20SLPM-D
COMPARE
±0.75% of reading ±0.1% of full scale 20 ±0.1% of reading ±0.02% of full scale
$1,970.00USD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter, 0 - 40 SLPM, Display, Battery EW-67803-30 Mfr # MWB-40SLPM-D
COMPARE
±0.75% of reading ±0.1% of full scale 40 ±0.1% of reading ±0.02% of full scale
$2,610.00USD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter, 0 - 50 SLPM, Display, Battery EW-67803-31 Mfr # MWB-50SLPM-D
COMPARE
±0.75% of reading ±0.1% of full scale 50 ±0.1% of reading ±0.02% of full scale
$2,610.00USD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter, 0 - 100 SLPM, Display, Battery EW-67803-32 Mfr # MWB-100SLPM-D
COMPARE
±0.75% of reading ±0.1% of full scale 100 ±0.1% of reading ±0.02% of full scale
$3,030.00USD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter, 0 - 250 SLPM, Display, Battery EW-67803-33 Mfr # MWB-250SLPM-D
COMPARE
±0.8% of reading ±0.2% of full scale 250 ±0.2% of reading ±0.02% of full scale
$3,030.00USD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter, 0 - 500 SLPM, Display, Battery EW-67803-34 Mfr # MWB-500SLPM-D
COMPARE
±0.8% of reading ±0.2% of full scale 500 ±0.2% of reading ±0.02% of full scale
$4,255.00USD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter, 0-1000 SLPM, Display, Battery EW-67803-35 Mfr # MWB-1000SLPM-D
COMPARE
±0.8% of reading ±0.2% of full scale 1000 ±0.2% of reading ±0.02% of full scale
$6,340.00USD / Each
Showing 1- 11 of 11