Amber and Clear Safety Bottles
Plain amber bottles (USP type 3) are available in 250-, 1000-, and 2500-mL sizes. PVC-coated amber bottle 07914-84 is designed for safety—won’t shatter like regular bottles. Clear, borosilicate glass bottle 07914-82 features a sidearm for a 0.2-µm air filter.
$43.20 - $79.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
DWK Life Sciences (Wheaton) 844025 Amber Glass Safety Bottle, USP Type 3, Square, 250 mL, 33 mm cap
Buy More Save More
EW-07914-80 Mfr # 844025
COMPARE
250 33 mm Glass
$43.20USD / Each
DWK Life Sciences (Wheaton) 844027 Amber Glass Safety Bottle, PVC-coated, 500 mL, GL32, 33 mm cap
Buy More Save More
EW-07914-84 Mfr # 844027
COMPARE
500 33 mm Glass
$79.50USD / Each
DWK Life Sciences (Wheaton) 844029 Amber Glass Safety Bottle, USP Type 3, Square, 1000 mL, 45 mm cap
Buy More Save More
EW-07914-86 Mfr # 844029
COMPARE
1000 45 mm Glass
$51.00USD / Each